Learn More
Biomatik Corporation Human Ribonuclease T2 (RNASET2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07128
Encompass_Preferred
Human Ribonuclease T2 (RNASET2) ELISA Kit for detecting RNASET2 in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O00584. Gene ID: 8635. Target Synonyms: Rnase-T2; RNASE6PL; RNASE6-PL; Ribonuclease 6. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3714