Learn More
Biomatik Corporation Human RNA Binding Motif Protein 20 (RBM20) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07141
Encompass_Preferred
Human RNA Binding Motif Protein 20 (RBM20) ELISA Kit for detecting RBM20 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.114ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q5T481. Gene ID: 282996. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3715