Learn More
Biomatik Corporation Human R-Spondin 1 (RSPO1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07145
Encompass_Preferred
Human R-Spondin 1 (RSPO1) ELISA Kit for detecting RSPO1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q2MKA7. Gene ID: 284654. Target Synonyms: RSPONDIN; Roof plate-specific spondin-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3716