Learn More
Biomatik Corporation Human R-Spondin 3 (RSPO3) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07147
Encompass_Preferred
Human R-Spondin 3 (RSPO3) ELISA Kit for detecting RSPO3 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.112ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9BXY4. Gene ID: 84870. Target Synonyms: CRISTIN1; PWTSR; THSD2; Thrombospondin,Type I,Domain Containing 2; Protein with TSP type-1 repeat; Roof plate-specific spondin-3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3718