Learn More
Biomatik Corporation Human Alpha-Fetoprotein Lens Culinaris Agglutiin 3, Afp-L3 Elisa Kit
Supplier:  Biomatik Corporation EKC40562
Encompass_Preferred
Human Alpha-Fetoprotein Lens Culinaris Agglutiin 3, Afp-L3 Elisa Kit for detecting Alpha-Fetoprotein Lens Culinaris Agglutiin 3, Afp-L3 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.3ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7755