Learn More
Biomatik Corporation Human Runt Related Transcription Factor 2 (RUNX2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07148
Encompass_Preferred
Human Runt Related Transcription Factor 2 (RUNX2) ELISA Kit for detecting RUNX2 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q13950. Gene ID: 860. Target Synonyms: AML3; CBFA1; CCD1; OSF2; PEBP2A; Acute myeloid leukemia 3 ; Core-binding factor alpha-1; Osteoblast-specific transcription factor 2; Polyomavirus enhancer-binding protein 2 A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3719