Learn More
Biomatik Corporation Human Ryanodine Receptor 1, Skeletal (RYR1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07150
Encompass_Preferred
Human Ryanodine Receptor 1, Skeletal (RYR1) ELISA Kit for detecting RYR1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.78-50ng/ml. Sensitivity: 0.30ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P21817. Gene ID: 6261. Target Synonyms: CCO; MHS; MHS1; RYDR; RYR; SKRR; Central Core Disease Of Muscle; Skeletal muscle calcium release channel. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3720