Learn More
Biomatik Corporation Dog Interleukin 4, Il-4 Elisa Kit
Supplier:  Biomatik Corporation EKC40791
Encompass_Preferred
Dog Interleukin 4, Il-4 Elisa Kit for detecting Interleukin 4 in serum, plasma, tissue homogenates. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 6.25-400pg/ml. Sensitivity: 1.56pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: O77762; IL4. Target Synonyms: IL4Interleukin-4; IL-4; B-cell stimulatory factor 1; BSF-1; Lymphocyte stimulatory factor 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7786