Learn More
Biomatik Corporation Rat Ryanodine Receptor 1, Skeletal (RYR1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07152
Encompass_Preferred
Rat Ryanodine Receptor 1, Skeletal (RYR1) ELISA Kit for detecting RYR1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.625-40ng/ml. Sensitivity: 0.231ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: F1LMY4. Target Synonyms: CCO; MHS; MHS1; RYDR; RYR; SKRR; Central Core Disease Of Muscle; Skeletal muscle calcium release channel. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3721