Learn More
Biomatik Corporation Human Growth Differentiation Factor-9, Gdf-9 Elisa Kit
Supplier:  Biomatik Corporation EKC40904
Encompass_Preferred
Human Growth Differentiation Factor-9, Gdf-9 Elisa Kit for detecting Growth Differentiation Factor 9 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 43-1500pg/ml. Sensitivity: 22pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: O60383; GDF9. Target Synonyms: OTTHUMP00000066213. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7796