Learn More
Biomatik Corporation Rat Carcinoembryonic Antign, Cea Elisa Kit
Supplier:  Biomatik Corporation EKC41081
Encompass_Preferred
Rat Carcinoembryonic Antign, Cea Elisa Kit for detecting Carcinoembryonic Antign, Cea in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7820