Learn More
Biomatik Corporation Human S100 Calcium Binding Protein (S100) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07155
Encompass_Preferred
Human S100 Calcium Binding Protein (S100) ELISA Kit for detecting S100 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.78-50ng/ml. Sensitivity: 0.27ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P23297. Gene ID: 6271. Target Synonyms: S100-A1; S100A1; S100-Alpha; S-100 Protein; S-100 protein subunit alpha; S-100 protein alpha chain. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3723