Learn More
Biomatik Corporation Guinea pig Substance P(SP)ELISA Kit
Supplier:  Biomatik Corporation EKC41265
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Cavia porcellus (Guinea pig). Detection Range: 47 pg/mL-3000 pg/mL. Sensitivity: 11.8 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-228-7839