Learn More
Biomatik Corporation Human S100 Calcium Binding Protein A11 (S100A11) ELISA Kit, 96 tests
Catalog No. 501503726
Encompass_Preferred
Human S100 Calcium Binding Protein A11 (S100A11) ELISA Kit for detecting S100A11 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.058ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P31949. Gene ID: 6282. Target Synonyms: S100-A11; S100C; MLN70; Protein S100-C; Calgizzarin; Metastatic lymph node gene 70 protein; Protein S100-A11, N-terminally processed. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3726 Supplier Biomatik Corporation Supplier No. EKU07161
Human S100 Calcium Binding Protein A11 (S100A11) ELISA Kit for detecting S100A11 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.058ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P31949. Gene ID: 6282. Target Synonyms: S100-A11; S100C; MLN70; Protein S100-C; Calgizzarin; Metastatic lymph node gene 70 protein; Protein S100-A11, N-terminally processed. Restrictions: For Research Use Only. Not for use in diagnostic procedures.