Learn More
Biomatik Corporation Human S100 Calcium Binding Protein A12 (S100A12) ELISA Kit, 96 tests
Catalog No. 501503727
Encompass_Preferred
Human S100 Calcium Binding Protein A12 (S100A12) ELISA Kit for detecting S100A12 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78-5,000pg/ml. Sensitivity: 33pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P80511. Gene ID: 6283. Target Synonyms: CGRP; P6; CAAF1; CAGC; ENRAGE; MRP6; Calgranulin C; Calcium-binding protein in amniotic fluid 1; Extracellular newly identified RAGE-binding; Migration inhibitory factor-related 6. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3727 Supplier Biomatik Corporation Supplier No. EKU07164
Human S100 Calcium Binding Protein A12 (S100A12) ELISA Kit for detecting S100A12 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78-5,000pg/ml. Sensitivity: 33pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P80511. Gene ID: 6283. Target Synonyms: CGRP; P6; CAAF1; CAGC; ENRAGE; MRP6; Calgranulin C; Calcium-binding protein in amniotic fluid 1; Extracellular newly identified RAGE-binding; Migration inhibitory factor-related 6. Restrictions: For Research Use Only. Not for use in diagnostic procedures.