Learn More
Biomatik Corporation Human DEFB1 (Beta-defensin 1) ELISA Kit
Supplier:  Biomatik Corporation EKF57248
Encompass_Preferred
Human DEFB1 (Beta-defensin 1) ELISA Kit for detecting DEFB1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P60022. Target Synonyms: DEFB1; Beta-defensin 1; BD1; HBD1; BD-1; hBD-1; Defensin Beta 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7897