Learn More
Biomatik Corporation Human S100 Calcium Binding Protein A4 (S100A4) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07168
Encompass_Preferred
Human S100 Calcium Binding Protein A4 (S100A4) ELISA Kit for detecting S100A4 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.78-50ng/ml. Sensitivity: 0.30ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P26447. Gene ID: 6275. Target Synonyms: S100-A4; MTS1; 18A2; 42A; CAPL; P9KA; PEL98; Calcium protein; Calvasculin; Metastasin; Protein Mts1; Placental calcium-binding protein; Murine placental homolog. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3729