Learn More
Biomatik Corporation Human PIGR (Polymeric Immunoglobulin Receptor/Membrane Secretory Component) ELISA Kit
Supplier:  Biomatik Corporation EKF57339
Encompass_Preferred
Human PIGR (Polymeric Immunoglobulin Receptor/Membrane Secretory Component) ELISA Kit for detecting PIGR in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P01833. Target Synonyms: PIGR; SC; Polymeric immunoglobulin receptor; Membrane Secretory Component. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7922