Learn More
Biomatik Corporation Human S100 Calcium Binding Protein A7 (S100A7) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07175
Encompass_Preferred
Human S100 Calcium Binding Protein A7 (S100A7) ELISA Kit for detecting S100A7 in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 125-8,000pg/ml. Sensitivity: 50pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P31151. Gene ID: 6278. Target Synonyms: S100-A7; PSOR1; S100A7c; Psoriasin 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3731