Learn More
Biomatik Corporation Human MASP1 (Mannan-binding lectin serine protease 1) ELISA Kit
Supplier:  Biomatik Corporation EKF57416
Encompass_Preferred
Human MASP1 (Mannan-binding lectin serine protease 1) ELISA Kit for detecting MASP1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 93.75-6000pg/ml. Sensitivity: 56.25pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P48740. Target Synonyms: MASP1; Mannan-binding lectin serine protease 1; Serine protease 5; Ra-reactive factor serine protease p100; RaRF; Mannose-binding protein-associated serine protease; Mannose-binding lectin-associated serine protease 1; MASP-1; Complement factor MASP-3; Complement-activating component of Ra-reactive factor; CRARF; CRARF1; PRSS5;. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7939