Learn More
Biomatik Corporation Mouse S100 Calcium Binding Protein A8 (S100A8) ELISA Kit, 96 tests
Catalog No. 501503733
Encompass_Preferred
Mouse S100 Calcium Binding Protein A8 (S100A8) ELISA Kit for detecting S100A8 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 7.8-500pg/ml. Sensitivity: 3.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P27005. Gene ID: 20201. Target Synonyms: MRP8; MIF; 60B8AG; CAGA; CFAG; CGLA; CP-10; L1Ag; NIF; P8; Calgranulin A; Cystic fibrosis antigen; Leukocyte L1 complex light chain; Migration inhibitory factor-related protein 8. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3733 Supplier Biomatik Corporation Supplier No. EKU07179
Mouse S100 Calcium Binding Protein A8 (S100A8) ELISA Kit for detecting S100A8 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 7.8-500pg/ml. Sensitivity: 3.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P27005. Gene ID: 20201. Target Synonyms: MRP8; MIF; 60B8AG; CAGA; CFAG; CGLA; CP-10; L1Ag; NIF; P8; Calgranulin A; Cystic fibrosis antigen; Leukocyte L1 complex light chain; Migration inhibitory factor-related protein 8. Restrictions: For Research Use Only. Not for use in diagnostic procedures.