Learn More
Biomatik Corporation Human S100 Calcium Binding Protein A9 (S100A9) ELISA Kit, 96 tests
Catalog No. 501503735
Encompass_Preferred
Human S100 Calcium Binding Protein A9 (S100A9) ELISA Kit for detecting S100A9 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.8pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P06702. Gene ID: 6280. Target Synonyms: MRP14; MIF; 60B8AG; CFAG; NIF; CAGB; CGLB; L1AG; P14; Calgranulin B; Migration inhibitory factor-related protein 14; Calprotectin L1H subunit; Leukocyte L1 complex heavy chain. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3735 Supplier Biomatik Corporation Supplier No. EKU07182
Human S100 Calcium Binding Protein A9 (S100A9) ELISA Kit for detecting S100A9 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.8pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P06702. Gene ID: 6280. Target Synonyms: MRP14; MIF; 60B8AG; CFAG; NIF; CAGB; CGLB; L1AG; P14; Calgranulin B; Migration inhibitory factor-related protein 14; Calprotectin L1H subunit; Leukocyte L1 complex heavy chain. Restrictions: For Research Use Only. Not for use in diagnostic procedures.