Learn More
Biomatik Corporation Human IFITM3 (Interferon-induced transmembrane protein 3) ELISA Kit
Supplier:  Biomatik Corporation EKF58009
Encompass_Preferred
Human IFITM3 (Interferon-induced transmembrane protein 3) ELISA Kit for detecting IFITM3 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q01628. Target Synonyms: IFITM3; Fragilis; IP15; mil-1; 1-8U; DSPA2b; interferon induced transmembrane protein 3; interferon-induced transmembrane protein 3; Interferon-inducible protein 1-8U. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8006