Learn More
Biomatik Corporation Mouse S100 Calcium Binding Protein B (S100B) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07185
Encompass_Preferred
Mouse S100 Calcium Binding Protein B (S100B) ELISA Kit for detecting S100B in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 12.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P50114. Gene ID: 20203. Target Synonyms: S100-B; NEF; S100Beta; S-100 protein subunit beta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3738