Learn More
Biomatik Corporation Human CML (Carboxymethyl Lysine) ELISA Kit
Supplier:  Biomatik Corporation EKF58037
Encompass_Preferred
Human CML (Carboxymethyl Lysine) ELISA Kit for detecting CML in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.625-1000ng/ml. Sensitivity: 9.375ng/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Target Synonyms: CML. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8018