Learn More
Biomatik Corporation Human S100 Calcium Binding Protein P (S100P) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07187
Encompass_Preferred
Human S100 Calcium Binding Protein P (S100P) ELISA Kit for detecting S100P in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.067ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P25815. Gene ID: 6286. Target Synonyms: S100-P; MIG9; S100-E; Migration-inducing gene 9 protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3740