Learn More
Biomatik Corporation Mouse calprotectin (S100A8/S100A9 Heterodimer) ELISA Kit
Supplier:  Biomatik Corporation EKF58285
Encompass_Preferred
Mouse calprotectin (S100A8/S100A9 Heterodimer) ELISA Kit for detecting Calprotectin in serum, plasma, cell culture supernatant, cell or tissue lysate, fecal extracts, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.25-80ng/ml. Sensitivity: 0.75ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P27005; P31725. Target Synonyms: S100 Calcium Binding Protein A8/S100 Calcium Binding Protein A9; Calprotectin; S100A8/S100A9 Heterodimer. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8086