Learn More
Biomatik Corporation Human Sclerostin (SOST) ELISA Kit, 96 tests
Catalog No. 501503744
Encompass_Preferred
Human Sclerostin (SOST) ELISA Kit for detecting SOST in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.131ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9BQB4. Gene ID: 50964. Target Synonyms: VBCH; Sclerosteosis. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3744 Supplier Biomatik Corporation Supplier No. EKU07202
Human Sclerostin (SOST) ELISA Kit for detecting SOST in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.131ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9BQB4. Gene ID: 50964. Target Synonyms: VBCH; Sclerosteosis. Restrictions: For Research Use Only. Not for use in diagnostic procedures.