Learn More
Biomatik Corporation Mouse Secondary Lymphoid Tissue Chemokine (SLC) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07208
Encompass_Preferred
Mouse Secondary Lymphoid Tissue Chemokine (SLC) ELISA Kit for detecting SLC in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 7.8-500pg/ml. Sensitivity: 3.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P86793. Gene ID: 100042493. Target Synonyms: CCL21; 6Ckine; CKb9; ECL; SCYA21; TCA4; ECL; Chemokine (C-C-Motif) Ligand 21; Beta Chemokine Exodus-2; Efficient Chemoattractant For Lymphocytes. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3746