Learn More
Biomatik Corporation Human Secreted Frizzled Related Protein 1 (SFRP1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07213
Encompass_Preferred
Human Secreted Frizzled Related Protein 1 (SFRP1) ELISA Kit for detecting SFRP1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8N474. Gene ID: 6422. Target Synonyms: FRP1; FRP; FRP-1; FrzA; SARP2; Secreted apoptosis-related protein 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3747