Learn More
Biomatik Corporation Mouse Secreted Frizzled Related Protein 1 (SFRP1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07214
Encompass_Preferred
Mouse Secreted Frizzled Related Protein 1 (SFRP1) ELISA Kit for detecting SFRP1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 12.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8C4U3. Gene ID: 20377. Target Synonyms: FRP1; FRP; FRP-1; FrzA; SARP2; Secreted apoptosis-related protein 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3748