Learn More
Biomatik Corporation Rat Hepc (Hepcidin) ELISA Kit
Supplier:  Biomatik Corporation EKF58556
Encompass_Preferred
Rat Hepc (Hepcidin) ELISA Kit for detecting Hepc in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q99MH3. Target Synonyms: Hepc; Hepcidin; HAMP; HFE2B; LEAP1; PLTR; LEAP-1; hepcidin antimicrobial peptide; Putative liver tumor regressor; Liver-expressed antimicrobial peptide 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8149