Learn More
Biomatik Corporation Rat GLUT4 (Glucose Transporter 4) ELISA Kit
Supplier:  Biomatik Corporation EKF58604
Encompass_Preferred
Rat GLUT4 (Glucose Transporter 4) ELISA Kit for detecting GLUT4 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.625-40ng/ml. Sensitivity: 0.375ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P19357. Target Synonyms: GLUT4; SLC2A4; Solute carrier family 2; facilitated glucose transporter member 4; Glucose transporter type 4; insulin-responsive; GLUT-4; GLUT-4; insulin-responsive glucose transporter type 4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8159