Learn More
Biomatik Corporation Human Secreted Frizzled Related Protein 2 (SFRP2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07216
Encompass_Preferred
Human Secreted Frizzled Related Protein 2 (SFRP2) ELISA Kit for detecting SFRP2 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q96HF1. Gene ID: 6423. Target Synonyms: FRP-2; SARP1; SDF-5; Secreted apoptosis-related protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3750