Learn More
Biomatik Corporation Human ADAM10 (Disintegrin and metalloproteinase domain-containing protein 10) ELISA Kit
Supplier:  Biomatik Corporation EKF58640
Encompass_Preferred
Human ADAM10 (Disintegrin and metalloproteinase domain-containing protein 10) ELISA Kit for detecting ADAM10 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 62.5-4000pg/ml. Sensitivity: 37.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: O14672. Target Synonyms: ADAM10; CD156c; Kuzbanian; MADM; a disintegrin and metalloprotease domain 10; AD10; ADAM 10; ADAM metallopeptidase domain 10; CD156c; CD156c antigen; CDw156; disintegrin and metalloproteinase domain-containing protein 10. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8165