Learn More
Biomatik Corporation Human Secreted Frizzled Related Protein 4 (SFRP4) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07219
Encompass_Preferred
Human Secreted Frizzled Related Protein 4 (SFRP4) ELISA Kit for detecting SFRP4 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 62.5-4000pg/ml. Sensitivity: 25.8pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q6FHJ7. Gene ID: 6424. Target Synonyms: FRP-4; FRPHE; Frizzled protein, human endometrium. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3751