Learn More
Biomatik Corporation Human Secretin (SCT) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07226
Encompass_Preferred
Human Secretin (SCT) ELISA Kit for detecting SCT in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 18.52-1,500pg/ml. Sensitivity: 6.44pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P09683. Gene ID: 6343. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3754