Learn More
Biomatik Corporation Mouse Secretin (SCT) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07227
Encompass_Preferred
Mouse Secretin (SCT) ELISA Kit for detecting SCT in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 12.35-1,000pg/ml. Sensitivity: 4.83pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: Q08535. Gene ID: 20287. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3755