Learn More
Biomatik Corporation Human Secretogranin II (SCG2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07229
Encompass_Preferred
Human Secretogranin II (SCG2) ELISA Kit for detecting SCG2 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.25-80ng/ml. Sensitivity: 0.51ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P13521. Gene ID: 7857. Target Synonyms: SN; SgII; CHGC; Chromogranin C; Secretoneurin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3756