Learn More
Biomatik Corporation Human Secretory Immunoglobulin A (sIgA) ELISA Kit, 96 tests
Catalog No. 501503757
Encompass_Preferred
Human Secretory Immunoglobulin A (sIgA) ELISA Kit for detecting sIgA in serum, plasma, saliva, stool, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.2-2,000pg/ml. Sensitivity: 15.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Target Synonyms: S-IgA. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3757 Supplier Biomatik Corporation Supplier No. EKU07235
Human Secretory Immunoglobulin A (sIgA) ELISA Kit for detecting sIgA in serum, plasma, saliva, stool, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.2-2,000pg/ml. Sensitivity: 15.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Target Synonyms: S-IgA. Restrictions: For Research Use Only. Not for use in diagnostic procedures.