Learn More
Biomatik Corporation Rat β-EP (Beta-Endorphin) ELISA Kit
Supplier:  Biomatik Corporation EKF58918
Encompass_Preferred
Rat β-EP (Beta-Endorphin) ELISA Kit for detecting β-EP in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Accession Number: P01194. Target Synonyms: β-EP; Beta-Endorphin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8208