Learn More
Biomatik Corporation Mouse Secretory Immunoglobulin A (sIgA) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07236
Encompass_Preferred
Mouse Secretory Immunoglobulin A (sIgA) ELISA Kit for detecting sIgA in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.625-40ng/ml. Sensitivity: 0.274ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Target Synonyms: S-IgA. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3758