Learn More
Biomatik Corporation Human PPARGC1A (Peroxisome prolifeRator-activated receptor gamma coactivator 1-alpha) ELISA Kit
Supplier:  Biomatik Corporation EKF58928
Encompass_Preferred
Human PPARGC1A (Peroxisome prolifeRator-activated receptor gamma coactivator 1-alpha) ELISA Kit for detecting PPARGC1A in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9UBK2. Target Synonyms: PPARGC1A; Peroxisome prolifeRator-activated receptor gamma coactivator 1-alpha; PGC-1-alpha; PPAR-gamma coactivator 1-alpha; PPARGC-1-alpha; Ligand effect modulator 6; LEM6; PGC1; PGC1A; PPARGC1; PGC-1v;. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8211