Learn More
Biomatik Corporation Mouse Secretory Leukocyte Peptidase Inhibitor (SLPI) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07240
Encompass_Preferred
Mouse Secretory Leukocyte Peptidase Inhibitor (SLPI) ELISA Kit for detecting SLPI in serum, plasma, tissue homogenates, cell lysates, urine, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P97430. Gene ID: 20568. Target Synonyms: ALK1; ALP; MPI; BLPI; HUSI; HUSI-I; WAP4; WFDC4; Antileukoproteinase; Seminal proteinase inhibitor; Mucus proteinase inhibitor; WAP four-disulfide core domain protein 4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3760