Learn More
Biomatik Corporation Human TTR (Transthyretin) ELISA Kit
Supplier:  Biomatik Corporation EKF58942
Encompass_Preferred
Human TTR (Transthyretin) ELISA Kit for detecting TTR in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 7.813-500pg/ml. Sensitivity: 4.688pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P02766. Target Synonyms: TTR; Transthyretin; PALB; TBPA; ATTR; Prealbumin; HsT2651; prealbumin; amyloidosis type I; thyroxine-binding prealbumin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8216