Learn More
Biomatik Corporation Human MUC2 (Mucin-2) ELISA Kit
Supplier:  Biomatik Corporation EKF58952
Encompass_Preferred
Human MUC2 (Mucin-2) ELISA Kit for detecting MUC2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q02817. Target Synonyms: MUC2; Mucin-2; MLP; MUC-2; SMUC. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8218