Learn More
Biomatik Corporation Mouse MSTN (MSTN/Growth differentiation factor 8) ELISA Kit
Supplier:  Biomatik Corporation EKF59001
Encompass_Preferred
Mouse MSTN (MSTN/Growth differentiation factor 8) ELISA Kit for detecting Mstn in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: O08689. Target Synonyms: Mstn; MSTN; Myostatin; GDF-8; GDF8growth differentiation factor 8; growth; differentiation factor 8; Myostatin; myostatin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8224