Learn More
Biomatik Corporation Mouse Selectin, Leukocyte (SELL) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07247
Encompass_Preferred
Mouse Selectin, Leukocyte (SELL) ELISA Kit for detecting SELL in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.625-40ng/ml. Sensitivity: 0.253ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P18337. Gene ID: 20343. Target Synonyms: CD62L; L-Selectin; SEL-L; LAM1; LECAM1; LNHR; LSEL; LYAM1; Leu-8; Lyam-1; PLNHR; TQ1; HLHRc; MEL14; Lymphocyte Adhesion Molecule 1; Lymph Node Homing Receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3764