Learn More
Biomatik Corporation Mouse PDGFD (Platelet Derived Growth Factor D) ELISA Kit
Supplier:  Biomatik Corporation EKF59048
Encompass_Preferred
Mouse PDGFD (Platelet Derived Growth Factor D) ELISA Kit for detecting PDGFD in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q925I7. Target Synonyms: PDGFD; Iris-expressed growth factor; PDGF-D; platelet derived growth factor D; SCDGF-BMGC26867; SCDGFBplatelet-derived growth factor D; spinal cord derived growth factor B; Spinal cord-derived growth factor B; spinal cord-derived growth factor-B. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8233