Learn More
Biomatik Corporation Pig Selectin, Platelet (SELP) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07251
Encompass_Preferred
Pig P-Selectin (SELP) ELISA Kit for detecting SELP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 6.25-400ng/ml. Sensitivity: 2.64ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q29097. Gene ID: 397150. Target Synonyms: CD62P; Selectin, Platelet; PSEL; GMP140; LECAM3; GRMP; GMRP; PADGEM; Leukocyte-endothelial cell adhesion molecule 3; Platelet activation dependent granule-external membrane. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3767